Learn More
Cayman Chemical Dihomo-γ-Linolenic Acid Metabolite MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502987672
Encompass_Preferred
A mixture that contains prostaglandin E1, 15-keto prostaglandin E1, 13,14-dihydro-15-keto prostaglandin E1, 13,14-dihydro-15-keto prostaglandin F1α, prostaglandin F1α, 8(S)-HETrE, 12(S)-HETrE, and 15(S)-HETrE
Catalog No. 50-298-7672 Supplier Cayman Chemical Supplier No. 222811 EA
A mixture that contains prostaglandin E1, 15-keto prostaglandin E1, 13,14-dihydro-15-keto prostaglandin E1, 13,14-dihydro-15-keto prostaglandin F1α, prostaglandin F1α, 8(S)-HETrE, 12(S)-HETrE, and 15(S)-HETrE