Learn More
Cayman Chemical Dihomo-γ-Linolenoyl Ethanolamide, Purity: ≥98%, CAS: 150314-34-4, 50 mg
SDP
Catalog No. 502895466
Encompass_Preferred
An endocannabinoid containing dihomo-γ-linoleate in place of the arachidonate moiety of AEA; binds to recombinant human CB1 and CB2 receptors with Ki values of 857 and 598 nM, respectively
Catalog No. 50-289-5466 Supplier Cayman Chemical Supplier No. 9023550 MG
An endocannabinoid containing dihomo-γ-linoleate in place of the arachidonate moiety of AEA; binds to recombinant human CB1 and CB2 receptors with Ki values of 857 and 598 nM, respectively