Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydralazine sulfate | 7327-87-9 | 10 MG
SDP

Catalog No. 5000419642
Encompass_Preferred

Dihydralazine sulfate is a vasodilator and an antihypertensive agent for research applications.

  • Purity: 98.0%
  • Molecular weight: 288.28
  • Molecular formula: C8H12N6O4S
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Soluble in DMSO (4.17 mg/mL)
  • Insoluble in water (< 0.1 mg/mL)

Catalog No. 50-004-19642 Supplier Medchemexpress LLC Supplier No. HYN706510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydralazine sulfate is a vasodilator and an antihypertensive agent for research applications.

  • Purity: 98.0%
  • Molecular weight: 288.28
  • Molecular formula: C8H12N6O4S
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Soluble in DMSO (4.17 mg/mL)
  • Insoluble in water (< 0.1 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.