Learn More
Medchemexpress LLC Dihydralazine sulfate | 7327-87-9 | 100 MG
SDP
Catalog No. 5000420128
Encompass_Preferred
Dihydralazine sulfate is a potent vasodilator and an effective antihypertensive agent, primarily used for research purposes. Studies have indicated its ability to inhibit hemorrhagic hypotension without negatively impacting cerebral blood flow in various hypertensive animal models.
  • Functions as a vasodilator
  • Acts as an antihypertensive agent
  • Intended for research use only
  • Shown to inhibit hemorrhagic hypotension in in vivo studies
  • Maintains cerebral blood flow during blood pressure reduction
Catalog No. 50-004-20128 Supplier Medchemexpress LLC Supplier No. HYN7065100MG
Dihydralazine sulfate is a potent vasodilator and an effective antihypertensive agent, primarily used for research purposes. Studies have indicated its ability to inhibit hemorrhagic hypotension without negatively impacting cerebral blood flow in various hypertensive animal models.
  • Functions as a vasodilator
  • Acts as an antihypertensive agent
  • Intended for research use only
  • Shown to inhibit hemorrhagic hypotension in in vivo studies
  • Maintains cerebral blood flow during blood pressure reduction