Learn More
Medchemexpress LLC Dihydralazine (sulfate) | 7327-87-9 | 25 MG
SDP
Catalog No. 5000421791
Encompass_Preferred
Dihydralazine sulfate is a vasodilator and an antihypertensive agent. In vivo studies have shown that it inhibits haemorrhagic hypotension in rats with renal hypertension (RHR) and spontaneous hypertension (SHR) without causing a concomitant fall in cerebral blood flow (CBF) when administered intravenously at single doses ranging from 0.1 to 2 mg/kg.
  • Acts as a vasodilator
  • Functions as an antihypertensive agent
  • Inhibits haemorrhagic hypotension in hypertensive rats without reducing cerebral blood flow
Catalog No. 50-004-21791 Supplier Medchemexpress LLC Supplier No. 25MGHYN7065
Dihydralazine sulfate is a vasodilator and an antihypertensive agent. In vivo studies have shown that it inhibits haemorrhagic hypotension in rats with renal hypertension (RHR) and spontaneous hypertension (SHR) without causing a concomitant fall in cerebral blood flow (CBF) when administered intravenously at single doses ranging from 0.1 to 2 mg/kg.
  • Acts as a vasodilator
  • Functions as an antihypertensive agent
  • Inhibits haemorrhagic hypotension in hypertensive rats without reducing cerebral blood flow