Learn More
Medchemexpress LLC Dihydralazine sulfate | 7327-87-9 | 5 MG
SDP
Catalog No. 5000419886
Encompass_Preferred
Dihydralazine sulfate is a chemical compound that acts as a vasodilator and an antihypertensive agent. It has been shown to inhibit hemorrhagic hypotension without a concomitant fall in cerebral blood flow in rats with renal hypertension and spontaneous hypertension. This product is intended for research applications only.
  • Acts as a vasodilator
  • Functions as an antihypertensive agent
  • Inhibits hemorrhagic hypotension
  • Suitable for research applications
Catalog No. 50-004-19886 Supplier Medchemexpress LLC Supplier No. HYN70655MG
Dihydralazine sulfate is a chemical compound that acts as a vasodilator and an antihypertensive agent. It has been shown to inhibit hemorrhagic hypotension without a concomitant fall in cerebral blood flow in rats with renal hypertension and spontaneous hypertension. This product is intended for research applications only.
  • Acts as a vasodilator
  • Functions as an antihypertensive agent
  • Inhibits hemorrhagic hypotension
  • Suitable for research applications