Learn More
Medchemexpress LLC Dihydralazine sulfate | 7327-87-9 | 50 MG
SDP
Catalog No. 5000420130
Encompass_Preferred
Dihydralazine sulfate is a vasodilator and an antihypertensive agent, intended for research use only. In vivo studies have shown that dihydralazine sulfate inhibits haemorrhagic hypotension without a concomitant fall in cerebral blood flow in hypertensive rats.
  • Vasodilator
  • Antihypertensive agent
  • For research use only
  • Inhibits haemorrhagic hypotension
  • Maintains cerebral blood flow in hypertensive rats
Catalog No. 50-004-20130 Supplier Medchemexpress LLC Supplier No. HYN706550MG
Dihydralazine sulfate is a vasodilator and an antihypertensive agent, intended for research use only. In vivo studies have shown that dihydralazine sulfate inhibits haemorrhagic hypotension without a concomitant fall in cerebral blood flow in hypertensive rats.
  • Vasodilator
  • Antihypertensive agent
  • For research use only
  • Inhibits haemorrhagic hypotension
  • Maintains cerebral blood flow in hypertensive rats