Learn More
Selleck Chemical LLC Dihydralazine sulphate S5030-1g
SDP
Catalog No. 503110913
Encompass_Preferred
Dihydralazine sulphate (Nepresol Depressan 1 4-Dihydrazinophthalazine Hydralazine) is a dihydralazine-l 4-phthalazine derivative and used as an antihypertensive drug
Catalog No. 50-311-0913 Supplier Selleck Chemical LLC Supplier No. S50301G
Dihydralazine sulphate (Nepresol Depressan 1 4-Dihydrazinophthalazine Hydralazine) is a dihydralazine-l 4-phthalazine derivative and used as an antihypertensive drug