Learn More
Selleck Chemical LLC Dihydralazine sulphate S5030-25mg
SDP
Catalog No. 503112453
Encompass_Preferred
Dihydralazine sulphate (Nepresol Depressan 1 4-Dihydrazinophthalazine Hydralazine) is a dihydralazine-l 4-phthalazine derivative and used as an antihypertensive drug
Catalog No. 50-311-2453 Supplier Selleck Chemical LLC Supplier No. S503025MG
Dihydralazine sulphate (Nepresol Depressan 1 4-Dihydrazinophthalazine Hydralazine) is a dihydralazine-l 4-phthalazine derivative and used as an antihypertensive drug