Learn More
Medchemexpress LLC HY-101299B 10mg Medchemexpress, Dihydrexidine (hydrochloride) CAS:137417-08-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101299B10MG
Encompass_Preferred
Medchemexpress, HY-101299B 10mg Dihydrexidine (HCl) CAS:137417-08-4 Dihydrexidine hydrochloride (DAR-100 hydrochloride) is a high potent, selective and full efficacy D1 dopamine receptor agonist with an IC50 of 10 nM, and displays some affinity for the D2 receptor. Dihydrexidine hydrochloride exhibits potent antiparkinsonian activity[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7827