Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydro dutasteride | 164656-22-8 | MFCD24386648 | 99.7% | 530.55 g/mol | C27H32F6N2O2 | 5 MG
SDP

Catalog No. 5000221820
Encompass_Preferred

Dihydro dutasteride is the active metabolite of dutasteride and acts as a potent inhibitor of 5α-reductase isozymes. Supplied as a high-purity research reference standard, it is intended for use in biochemical studies, metabolite profiling, assay development, and mechanism-of-action experiments.

  • High purity suitable for analytical and reference use.
  • Molecular weight 530.55 g/mol.
  • Chemical formula C27H32F6N2O2.
  • Available in small pack sizes for laboratory assays.
  • Used as a reference standard in metabolism and enzyme inhibition studies.

Catalog No. 50-002-21820 Supplier Medchemexpress LLC Supplier No. HY1353855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydro dutasteride is the active metabolite of dutasteride and acts as a potent inhibitor of 5α-reductase isozymes. Supplied as a high-purity research reference standard, it is intended for use in biochemical studies, metabolite profiling, assay development, and mechanism-of-action experiments.

  • High purity suitable for analytical and reference use.
  • Molecular weight 530.55 g/mol.
  • Chemical formula C27H32F6N2O2.
  • Available in small pack sizes for laboratory assays.
  • Used as a reference standard in metabolism and enzyme inhibition studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.