Learn More
Medchemexpress LLC Dihydro-β-erythroidine hydrobromide | 29734-68-7 | >98.0% | 356.25 g·mol⁻¹ | C16H22BrNO3 | 5 MG
SDP
Catalog No. 5000240311
Encompass_Preferred
Dihydro-β-erythroidine hydrobromide is a research reagent that acts as a competitive antagonist of neuronal nicotinic acetylcholine receptors, with preferential activity at α4β2 and α4β4 subtypes. Supplied as the hydrobromide salt, it is used in electrophysiology, receptor pharmacology, and in vivo studies to selectively inhibit nAChR-mediated responses.
  • High purity (≥98% by HPLC).
  • Selective antagonist for α4β2 and α4β4 nicotinic receptors.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Supplied as hydrobromide salt for improved stability and solubility.
  • Available in small research package sizes (1 mg, 5 mg, 10 mg).
  • Documented identifiers include CAS 29734-68-7 and molecular weight 356.25 g·mol⁻¹.
Catalog No. 50-002-40311 Supplier Medchemexpress LLC Supplier No. HY1076705MG
Dihydro-β-erythroidine hydrobromide is a research reagent that acts as a competitive antagonist of neuronal nicotinic acetylcholine receptors, with preferential activity at α4β2 and α4β4 subtypes. Supplied as the hydrobromide salt, it is used in electrophysiology, receptor pharmacology, and in vivo studies to selectively inhibit nAChR-mediated responses.
  • High purity (≥98% by HPLC).
  • Selective antagonist for α4β2 and α4β4 nicotinic receptors.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Supplied as hydrobromide salt for improved stability and solubility.
  • Available in small research package sizes (1 mg, 5 mg, 10 mg).
  • Documented identifiers include CAS 29734-68-7 and molecular weight 356.25 g·mol⁻¹.