Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydro-β-erythroidine hydrobromide | 29734-68-7 | MFCD00153793 | >98.0% | 356.25 g/mol | C16H22BrNO3 | 10 MG
SDP

Catalog No. 5000241292
Encompass_Preferred

Dihydro-β-erythroidine hydrobromide is a competitive antagonist of neuronal nicotinic acetylcholine receptors (nAChRs), with selectivity for α4β2 and α4β4 subtypes. It is used as a pharmacological tool in receptor characterization, electrophysiology, and behavioral studies to probe cholinergic signaling.

  • Competitive nAChR antagonist selective for α4β2 and α4β4.
  • CAS number 29734-68-7.
  • Chemical formula C16H22BrNO3; molecular weight 356.25 g/mol.
  • Purity >98.0% (HPLC) as reported by catalog vendors.
  • Typical research pack sizes: 1 mg, 5 mg, 10 mg.
  • Common applications: electrophysiology, receptor pharmacology, and behavioral assays.

Catalog No. 50-002-41292 Supplier Medchemexpress LLC Supplier No. HY10767010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydro-β-erythroidine hydrobromide is a competitive antagonist of neuronal nicotinic acetylcholine receptors (nAChRs), with selectivity for α4β2 and α4β4 subtypes. It is used as a pharmacological tool in receptor characterization, electrophysiology, and behavioral studies to probe cholinergic signaling.

  • Competitive nAChR antagonist selective for α4β2 and α4β4.
  • CAS number 29734-68-7.
  • Chemical formula C16H22BrNO3; molecular weight 356.25 g/mol.
  • Purity >98.0% (HPLC) as reported by catalog vendors.
  • Typical research pack sizes: 1 mg, 5 mg, 10 mg.
  • Common applications: electrophysiology, receptor pharmacology, and behavioral assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.