Learn More
AdipoGen D-erythro-Dihydrosphingosine 1-phosphate (1 mg)
SDP
Catalog No. 502053750
Encompass_Preferred
Chemical. CAS: 19794-97-9. Formula: C18H40NO5P. MW: 381.5. Saturated analog of sphingosine 1-phosphate (1-SP1). Ligand for sphingosine 1-phosphate (S1P/EDG) receptors. Negative control for intracellular effects of sphingosine 1-phosphate. Induces chemotaxis. Antifibrotic.
Catalog No. 50-205-3750 Supplier AdipoGen Supplier No. AGCR10005M001
Chemical. CAS: 19794-97-9. Formula: C18H40NO5P. MW: 381.5. Saturated analog of sphingosine 1-phosphate (1-SP1). Ligand for sphingosine 1-phosphate (S1P/EDG) receptors. Negative control for intracellular effects of sphingosine 1-phosphate. Induces chemotaxis. Antifibrotic.