Learn More
Medchemexpress LLC HY-W041301 500mg Medchemexpress, (±)-Dihydroactinidiolide CAS:15356-74-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW041301500MG
Encompass_Preferred
Medchemexpress, HY-W041301 500mg (±)-Dihydroactinidiolide CAS:15356-74-8 (±)-Dihydroactinidiolide, an important aroma compound of black tea and tobacco, has been isolated from several plants. (±)-Dihydroactinidiolide can be formation from β-Carotene by the treatment of polyphenoloxidase, the lipoxygenase, and the xanthine oxidase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3475