Learn More
Cayman Chemical Dihydroartemisinic Acid, Purity: ≥98%, CAS: 85031-59-0, 10 mg
SDP
Catalog No. 502932466
Encompass_Preferred
A biosynthetic precursor to artemisinin; an intermediate in the synthesis of artemisinin from artemisinic acid
Catalog No. 50-293-2466 Supplier Cayman Chemical Supplier No. 2514210 MG
A biosynthetic precursor to artemisinin; an intermediate in the synthesis of artemisinin from artemisinic acid