Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydroartemisinic acid | 85031-59-0 | 1 ML
SDP

Catalog No. 5000420844
Encompass_Preferred

Dihydroartemisinic acid (Dihydroqinghao acid) is a biosynthetic precursor to the antimalarial agent Artemisinin. This compound is intended for research use only.

  • Biosynthetic precursor to antimalarial agent Artemisinin
  • Target: Plasmodium
  • Target: Ferroptosis
  • Pathway: Anti-infection
  • Pathway: Apoptosis
  • Purity: 99.58%
  • Molecular Weight: 236.35
  • Molecular Formula: C15H24O2
  • Appearance: White to off-white solid

Catalog No. 50-004-20844 Supplier Medchemexpress LLC Supplier No. HYN410610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydroartemisinic acid (Dihydroqinghao acid) is a biosynthetic precursor to the antimalarial agent Artemisinin. This compound is intended for research use only.

  • Biosynthetic precursor to antimalarial agent Artemisinin
  • Target: Plasmodium
  • Target: Ferroptosis
  • Pathway: Anti-infection
  • Pathway: Apoptosis
  • Purity: 99.58%
  • Molecular Weight: 236.35
  • Molecular Formula: C15H24O2
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.