Learn More
AdipoGen Dihydroartemisinin (1 g)
SDP
Supplier:  AdipoGen AGCN20468G001
Encompass_Preferred
Chemical. CAS: 71939-50-9. Formula: C15H24O5. MW: 284.3. Isolated from Artemisia annua. Active metabolite of artemisinin. Anti-malaria (Plasmodium). Anti-schistosoma. Apoptosis inducer. Anti-cancer. Anti-inflammatory Inhibits production of tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6), monocyte chemotactic protein-1 (MCP-1) and nitric oxide (NO). Autophagy inducer.
Catalog No. 50-205-3645