Learn More
Medchemexpress LLC Dihydroartemisinin | 71939-50-9 | 284.35 | 1 G
SDP
Catalog No. 5000427886
Encompass_Preferred
Dihydroartemisinin (DHA) is a potent antimalarial agent that modulates cellular processes. It up-regulates cytosolic RelA/p65 protein levels while down-regulating nuclear RelA/p65, effectively blocking its nuclear translocation. DHA also induces autophagy and suppresses NF-kB activation in RPMI 8226 cells. Furthermore, it can enhance the anti-tumor effect of photodynamic therapy (PDT) on esophageal cancer cells.
  • Potent anti-malaria activity
  • Modulates RelA/p65 protein levels
  • Blocks nuclear translocation of RelA/p65
  • Induces autophagy
  • Suppresses NF-kB activation
  • Enhances anti-tumor effects of photodynamic therapy
  • Derived from Artemisia carvifolia Buch.-Ham. ex Roxb. plants
Catalog No. 50-004-27886 Supplier Medchemexpress LLC Supplier No. HYN01761G
Dihydroartemisinin (DHA) is a potent antimalarial agent that modulates cellular processes. It up-regulates cytosolic RelA/p65 protein levels while down-regulating nuclear RelA/p65, effectively blocking its nuclear translocation. DHA also induces autophagy and suppresses NF-kB activation in RPMI 8226 cells. Furthermore, it can enhance the anti-tumor effect of photodynamic therapy (PDT) on esophageal cancer cells.
  • Potent anti-malaria activity
  • Modulates RelA/p65 protein levels
  • Blocks nuclear translocation of RelA/p65
  • Induces autophagy
  • Suppresses NF-kB activation
  • Enhances anti-tumor effects of photodynamic therapy
  • Derived from Artemisia carvifolia Buch.-Ham. ex Roxb. plants