Learn More
Abcam Dihydroartemisinin, active metabolite of artemether and artesunate, 100MG
Catalog No. 502706209
Encompass_Preferred
MW 284.35 Da. Induces cell cycle arrest and inhibits cell migration in cancer cells. Depletes cellular iron via transferrin receptor 1. Antimalarial and antitumor agent. Orally active. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6209 Supplier Abcam Supplier No. AB142690100MG
MW 284.35 Da. Induces cell cycle arrest and inhibits cell migration in cancer cells. Depletes cellular iron via transferrin receptor 1. Antimalarial and antitumor agent. Orally active. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement