Learn More
Abcam Dihydroartemisinin, active metabolite of artemether and artesunate, 50MG
Catalog No. 502706207
Encompass_Preferred
MW 284.35 Da. Induces cell cycle arrest and inhibits cell migration in cancer cells. Depletes cellular iron via transferrin receptor 1. Antimalarial and antitumor agent. Orally active. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6207 Supplier Abcam Supplier No. AB14269050MG
MW 284.35 Da. Induces cell cycle arrest and inhibits cell migration in cancer cells. Depletes cellular iron via transferrin receptor 1. Antimalarial and antitumor agent. Orally active. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement