Learn More
Selleck Chemical LLC Dihydroartemisinin (DHA) 100mg 71939-50-9
SDP
Catalog No. 505831
Encompass_Preferred
Dihydroartemisinin (DHA) is a semi-synthetic derivative of artemisinin and isolated from the traditional Chinese herb Artemisia annua. Dihydroartemisinin induces autophagy and apoptosis by suppressing NF-?B activation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-583-1 Supplier Selleck Chemical LLC Supplier No. S2290100MG
Dihydroartemisinin (DHA) is a semi-synthetic derivative of artemisinin and isolated from the traditional Chinese herb Artemisia annua. Dihydroartemisinin induces autophagy and apoptosis by suppressing NF-?B activation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.