Learn More
Medchemexpress LLC Dihydroartemisinin (mixture of α and β isomers) | 71939-50-9 | MFCD00274495 | 98.0% | 284.35 g/mol | C15H24O5 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW129233500MG
Encompass_Preferred
Dihydroartemisinin is an active metabolite of artemisinin used in research as an antimalarial and as a probe for antiproliferative and autophagy-related pathways. It is provided as a high-purity compound and is available in multiple solid and solution formats for in vitro studies.
  • High purity (98.0%) suitable for research applications.
  • Available as solids and as 10 mM solutions in DMSO for immediate use.
  • Relevant for studies of parasite biology, apoptosis, and autophagy.
  • Well-characterized with CAS number 71939-50-9 and published spectral data.
  • Supplied in small pack sizes convenient for bench-top experiments.
Catalog No. 50-002-83649