Learn More
Cayman Chemical Dihydroartemisinin (mixed isomers), Purity: ≥98%, CAS: 131175-87-6, 10 g
SDP
Catalog No. 502943870
Encompass_Preferred
A mixture of dihydroartemisinin isomers; contains α-dihydroartemisinin and β-dihydroartemisinin
Catalog No. 50-294-3870 Supplier Cayman Chemical Supplier No. 4190810 G
A mixture of dihydroartemisinin isomers; contains α-dihydroartemisinin and β-dihydroartemisinin