Learn More
Selleck Chemical LLC Dihydrocaffeic acid
SDP
Catalog No. 5000002966
Encompass_Preferred
3 4-Dihydroxyhydrocinnamic acid (Dihydrocaffeic acid) is a metabolite product of the hydrogenation of caffeoylquinic acids occurring in normal human biofluids with potent antioxidant properties
Catalog No. 50-000-02966 Supplier Selleck Chemical LLC Supplier No. S633825MG
3 4-Dihydroxyhydrocinnamic acid (Dihydrocaffeic acid) is a metabolite product of the hydrogenation of caffeoylquinic acids occurring in normal human biofluids with potent antioxidant properties