Learn More
AdipoGen Dihydrocapsaicin (25 mg)
SDP
Supplier:  AdipoGen CDXD0679M025
Encompass_Preferred
Chemical. CAS: 19408-84-5. Formula: C18H29NO3. MW: 307.43. Isolated from plant source. Dihydrocapsaicin is a capsaicinoid and analog and congener of capsaicin in chili peppers (capsicum). Dihydrocapsaicin accounts for about 10% of the total capsaicinoid mixture and separation by HPLC is required in order to obtain pure dihydrocapsaicin. Dihydrocapsaicin, similar as capsaicin elicits a sensation of burning pain by activation of VR1 on small, non-myelinated polymodal C-type nociceptive nerve fibers. VR1 (vanilloid receptor 1) is a heat activated calcium ion channel which functions as a part of the normal nociceptive pain pathway. Shown to be analgesic against arthritis pain and inflammation and protective effects against high cholesterol levels and obesity. Induced autophagy, apoptosis in glioma cells via ROS-dependent pathways.
Catalog No. 50-205-4396