Learn More
Enzo Life Sciences Dihydrocapsaicin (50mg). CAS: 19408-84-5
SDP
Supplier:  Enzo Life Sciences ALX350052M050
Encompass_Preferred
Dihydro-analog and congener of capsaicin (BML-EI125) in chili peppers (Capsicum). Like capsaicin it is irritant. Dihydrocapsaicin accounts for about 22% of the total capsaicinoids mixture and has about the same pungency as capsaicin. Antioxidant. Reduces oxidation of serum lipids. Mutagenic. Alternative name: 8-Methyl-N-vanillylnonanamide. Purity: ≥85% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in chloroform, DMSO or 100% ethanol; sparingly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8709