Learn More
Medchemexpress LLC Dihydrocapsaicin-d3 | 1330261-21-6 | 310.45 | 1 MG
SDP
Catalog No. 5000389342
Encompass_Preferred
Dihydrocapsaicin-d3 is the deuterium-labeled version of Dihydrocapsaicin. It is a capsaicin and a potent, selective TRPV1 (transient receptor potential vanilloid channel 1) agonist. This compound reduces AIF, Bax, and Caspase-3 expressions, while increasing Bcl-2, Bcl-xL, and p-Akt levels. It also enhances hypothermia-induced neuroprotection after ischemic stroke via PI3K/Akt regulation in rats.
  • Deuterium-labeled version of Dihydrocapsaicin
  • Potent, selective TRPV1 agonist
  • Reduces AIF, Bax, and Caspase-3 expressions
  • Increases Bcl-2, Bcl-xL, and p-Akt levels
  • Enhances hypothermia-induced neuroprotection after ischemic stroke via PI3K/Akt regulation in rats
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89342 Supplier Medchemexpress LLC Supplier No. HYN0361S1MG
Dihydrocapsaicin-d3 is the deuterium-labeled version of Dihydrocapsaicin. It is a capsaicin and a potent, selective TRPV1 (transient receptor potential vanilloid channel 1) agonist. This compound reduces AIF, Bax, and Caspase-3 expressions, while increasing Bcl-2, Bcl-xL, and p-Akt levels. It also enhances hypothermia-induced neuroprotection after ischemic stroke via PI3K/Akt regulation in rats.
  • Deuterium-labeled version of Dihydrocapsaicin
  • Potent, selective TRPV1 agonist
  • Reduces AIF, Bax, and Caspase-3 expressions
  • Increases Bcl-2, Bcl-xL, and p-Akt levels
  • Enhances hypothermia-induced neuroprotection after ischemic stroke via PI3K/Akt regulation in rats
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS