Learn More
AdipoGen Dihydrocapsiate
SDP
Supplier:  AdipoGen CDXD0419M100
Encompass_Preferred
Chemical. CAS 205687-03-2. Formula C18H28O4. MW 308.41. Synthetic. Structurally related to capsaicin and found in the non-pungent pepper strain, CH-19 Sweet. Activates the transient receptor potential vanilloid 1 TRPV1. Shown to have antioxidant, apoptosis incuding in tumor cell lines and anti-inflammatory activities. Inhibited vascular endothelial growth factor VEGF-induced proliferation, chemotactic motility, and capillary-like tube formation of primary cultured human endothelial cells. Stimulation of TRPV1 receptors in the gut are proposed to activate the SNS, which can increase lipogenesis and thermogenesis.
Catalog No. 50-256-1083