Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Dihydrocarvone

Catalog No. 3727525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-275-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-275-25ML Supplier Sigma Aldrich Supplier No. 3727525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-Dihydrocarvone, a monoterpenoid compound found in caraway oil, is a key building block to synthesize sesquiterpenes. It is generally produced either by the hydrogenation of carvone or oxidation of limonene.(+)-Dihydrocarvone may be used in the following processes:Synthesis of dispiro 1,2,4,5-tetraoxanes, which show potent anti-malarial activity.Synthesis of an epoxylactone by oxidation, which can undergo copolymerization with +a-caprolactone to form cross-linked copolymers with shape memory properties.Synthesis of +a-Cyperone, a eudesmane type sesquiterpenoid compound with potent insecticidal activity.

Specifications

CAS 18031-40-8
Density 0.965 g/mL at 25°C
Boiling Point 104°C to 105°C (10 mmHg, lit.)
Molecular Formula C10H14O
Refractive Index n20/D 1.471
Linear Formula C10H14O
Quantity 25 mL
MDL Number MFCD00001543
Synonym (2R,5R)-5-Isopropenyl-2-methylcyclohexanone
Molecular Weight (g/mol) 150.22 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.