Learn More
Medchemexpress LLC (+)-Dihydrocarvone | 5524-05-0 | 100 G
SDP
Catalog No. 5000417440
Encompass_Preferred
(+)-Dihydrocarvone is a chemical substance known for eliciting strong antennal responses in *Apis mellifera* and influencing bee foraging preference for tomato.
  • Elicits strong antennal responses of Apis mellifera
  • Influences bee foraging preference for tomato
  • Related to tomato flower volatiles
  • Acts as an inhibitor
Catalog No. 50-004-17440 Supplier Medchemexpress LLC Supplier No. HYN8491A100G
(+)-Dihydrocarvone is a chemical substance known for eliciting strong antennal responses in *Apis mellifera* and influencing bee foraging preference for tomato.
  • Elicits strong antennal responses of Apis mellifera
  • Influences bee foraging preference for tomato
  • Related to tomato flower volatiles
  • Acts as an inhibitor