Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Dihydrocarvone | 5524-05-0 | 50 G
SDP

Catalog No. 5000417367
Encompass_Preferred

(+)-Dihydrocarvone is a colorless to light yellow liquid derived from plants, with molecular weight 152.23 and formula C10H16O. This terpenoid and sesquiterpene boasts 98.92% purity. For research use only, it elicits strong antennal responses in *A. mellifera* and influences bee foraging preferences for tomato.

  • Purity: 98.92%
  • Molecular weight: 152.23
  • Molecular formula: C10H16O
  • Appearance: Colorless to light yellow liquid
  • Density: 0.904±0.06 g/cm3
  • Classification: Terpenoid, sesquiterpene from plants
  • Shipping: Room temperature
  • Storage: -20°C (under nitrogen); in solvent: -80°C (6 months), -20°C (1 month)
  • Biological activity: Elicits strong antennal responses in A. mellifera

Catalog No. 50-004-17367 Supplier Medchemexpress LLC Supplier No. HYN8491A50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Dihydrocarvone is a colorless to light yellow liquid derived from plants, with molecular weight 152.23 and formula C10H16O. This terpenoid and sesquiterpene boasts 98.92% purity. For research use only, it elicits strong antennal responses in *A. mellifera* and influences bee foraging preferences for tomato.

  • Purity: 98.92%
  • Molecular weight: 152.23
  • Molecular formula: C10H16O
  • Appearance: Colorless to light yellow liquid
  • Density: 0.904±0.06 g/cm3
  • Classification: Terpenoid, sesquiterpene from plants
  • Shipping: Room temperature
  • Storage: -20°C (under nitrogen); in solvent: -80°C (6 months), -20°C (1 month)
  • Biological activity: Elicits strong antennal responses in A. mellifera

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.