Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydrochalcone 10g | 1083-30-3 | 10 G
SDP

Catalog No. 5000307589
Encompass_Preferred

Dihydrochalcone is a small-molecule research reagent reported to inhibit Aβ aggregation and to penetrate the blood-brain barrier. Supplied as a solid with high reported purity, it is intended for biochemical assays and neuroscience research.

  • High reported purity: 99.87%.
  • Molecular weight 210.27 g/mol.
  • Chemical formula C15H14O.
  • Available in laboratory pack sizes including 10 g.
  • Suitable for Aβ aggregation and biochemical assays.

Catalog No. 50-003-07589 Supplier Medchemexpress LLC Supplier No. HY11450810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrochalcone is a small-molecule research reagent reported to inhibit Aβ aggregation and to penetrate the blood-brain barrier. Supplied as a solid with high reported purity, it is intended for biochemical assays and neuroscience research.

  • High reported purity: 99.87%.
  • Molecular weight 210.27 g/mol.
  • Chemical formula C15H14O.
  • Available in laboratory pack sizes including 10 g.
  • Suitable for Aβ aggregation and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.