Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydrochalcone 25g | 1083-30-3 | 210.27 g/mol | C15H14O | 25 G
SDP

Catalog No. 5000229590
Encompass_Preferred

Dihydrochalcone (CAS 1083-30-3) is a small-molecule research reagent supplied as a high-purity solid for in vitro and analytical applications.

  • High purity ≥98%.
  • Molecular formula C15H14O.
  • Molecular weight 210.27 g/mol.
  • Suitable for in vitro biochemical assays and as an analytical reference.
  • Available in a 25 G pack for laboratory use.

Catalog No. 50-002-29590 Supplier Medchemexpress LLC Supplier No. HY11450825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrochalcone (CAS 1083-30-3) is a small-molecule research reagent supplied as a high-purity solid for in vitro and analytical applications.

  • High purity ≥98%.
  • Molecular formula C15H14O.
  • Molecular weight 210.27 g/mol.
  • Suitable for in vitro biochemical assays and as an analytical reference.
  • Available in a 25 G pack for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.