Learn More
Cayman Chemical Dihydrochelerythrine, Purity: ≥95%, CAS: 6880-91-7, 10 mg
SDP
Catalog No. 502991394
Encompass_Preferred
An alkaloid with diverse biological activities; active against S. aureus, S. faecalis, and C. albicans (MIC = 1.87 μg/ml for all); inhibits secretion of the HBV antigens HBsAg and HBeAg from HepG2 2.2.15 cells (IC50s = 50s = 1.4, 0.4, and 3.5 μM, respectively)
Catalog No. 50-299-1394 Supplier Cayman Chemical Supplier No. 3011810 MG
An alkaloid with diverse biological activities; active against S. aureus, S. faecalis, and C. albicans (MIC = 1.87 μg/ml for all); inhibits secretion of the HBV antigens HBsAg and HBeAg from HepG2 2.2.15 cells (IC50s = 50s = 1.4, 0.4, and 3.5 μM, respectively)