Learn More
AdipoGen Dihydrochlamydocin
SDP
Supplier:  AdipoGen AGCN20115M001
Encompass_Preferred
Chemical. CAS 157618-75-2. Formula C28H40N4O6. MW 528.7. Isolated from Verticillium sp. Phytotoxin. Derivative of HDAC inhibitor chlamydocin Similar chemical structure to the TAN-1746 compounds potent HDAC inhibitors.
Catalog No. 50-256-2656