Learn More
Medchemexpress LLC Dihydroconiferyl alcohol | 2305-13-7 | MFCD00016571 | 99.9% | 182.22 g/mol | C10H14O3 | 1 ML
SDP
Catalog No. 5000302242
Encompass_Preferred
Dihydroconiferyl alcohol is provided as a ready-to-use 10 mM solution in DMSO (1 mL) for research applications as a cell division factor and endogenous metabolite probe (CAS 2305-13-7). The product is supplied with technical and safety documentation and should be handled and stored protected from light.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.9%) for reproducible experimental results.
  • Suitable for cell division and metabolite-related assays.
  • Store protected from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Includes safety data sheet and product data sheet for handling and storage.
Catalog No. 50-003-02242 Supplier Medchemexpress LLC Supplier No. HYW01639310MM/1ML
Dihydroconiferyl alcohol is provided as a ready-to-use 10 mM solution in DMSO (1 mL) for research applications as a cell division factor and endogenous metabolite probe (CAS 2305-13-7). The product is supplied with technical and safety documentation and should be handled and stored protected from light.
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.9%) for reproducible experimental results.
  • Suitable for cell division and metabolite-related assays.
  • Store protected from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Includes safety data sheet and product data sheet for handling and storage.