Learn More
Medchemexpress LLC Dihydrocoumarin (Hydrocoumarin) | 119-84-6 | 148.16 | 50 G
SDP
Catalog No. 5000415397
Encompass_Preferred
Dihydrocoumarin is a compound found in Melilotus officinalis. It is a yeast Sir2p inhibitor and also inhibits human SIRT1 and SIRT2 with IC50s of 208 μM and 295 μM, respectively.
  • Found in Melilotus officinalis.
  • Yeast Sir2p inhibitor.
  • Inhibits human SIRT1 (IC50 of 208 μM) and SIRT2 (IC50 of 295 μM).
  • Induces concentration-dependent inhibition of SIRT1 in an in vitro enzymatic assay.
  • Increases cytotoxicity and apoptosis in the TK6 cell line in a dose-dependent manner.
  • Increases p53 lysine 373 and 382 acetylation in the TK6 cell line.
Catalog No. 50-004-15397 Supplier Medchemexpress LLC Supplier No. HYN192650G
Dihydrocoumarin is a compound found in Melilotus officinalis. It is a yeast Sir2p inhibitor and also inhibits human SIRT1 and SIRT2 with IC50s of 208 μM and 295 μM, respectively.
  • Found in Melilotus officinalis.
  • Yeast Sir2p inhibitor.
  • Inhibits human SIRT1 (IC50 of 208 μM) and SIRT2 (IC50 of 295 μM).
  • Induces concentration-dependent inhibition of SIRT1 in an in vitro enzymatic assay.
  • Increases cytotoxicity and apoptosis in the TK6 cell line in a dose-dependent manner.
  • Increases p53 lysine 373 and 382 acetylation in the TK6 cell line.