Learn More
Cayman Chemical DIhydrocurcumIn 10mg
SDP
Catalog No. 502943509
Encompass_Preferred
An active metabolite of curcumin; reduces triglyceride levels and increases glucose uptake in HepG2 and L-02 cellular models of oleic acid-induced steatosis at 20 µM
Catalog No. 50-294-3509 Supplier Cayman Chemical Supplier No. 3982310 MG
An active metabolite of curcumin; reduces triglyceride levels and increases glucose uptake in HepG2 and L-02 cellular models of oleic acid-induced steatosis at 20 µM