Learn More
Medchemexpress LLC HY-N1967 10mg Medchemexpress, Dihydrocurcumin CAS:76474-56-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN196710MG
Encompass_Preferred
Medchemexpress, HY-N1967 10mg Dihydrocurcumin CAS:76474-56-1 Dihydrocurcumin, a major metabolites of curcumin, reduces lipid accumulation and oxidative stress. Dihydrocurcumin regulates mRNA and protein expression levels of SREBP-1C, PNPLA3 and PPARα, increases protein expression levels of pAKT and PI3K, and reduced the levels of cellular NO and ROS via Nrf2 signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2854