Learn More
Cayman Chemical DIhydrodaIdzeIn 10mg
SDP
Catalog No. 502968283
Encompass_Preferred
An active, estrogenic metabolite of daidzein; has vasodilatory action on rat isolated aortic rings at 1 UG/ml; stimulates the estrogen receptor-dependent growth of breast cancer MCF-7 cells at micromolar concentrations
Catalog No. 50-296-8283 Supplier Cayman Chemical Supplier No. 1742310 MG
An active, estrogenic metabolite of daidzein; has vasodilatory action on rat isolated aortic rings at 1 UG/ml; stimulates the estrogen receptor-dependent growth of breast cancer MCF-7 cells at micromolar concentrations