Learn More
Cayman Chemical Dihydrodaidzein, Purity: ≥98%, CAS: 17238-05-0, 50 mg
SDP
Catalog No. 502967370
Encompass_Preferred
An active, estrogenic metabolite of daidzein; has vasodilatory action on rat isolated aortic rings at 1 µg/ml; stimulates the estrogen receptor-dependent growth of breast cancer MCF-7 cells at micromolar concentrations
Catalog No. 50-296-7370 Supplier Cayman Chemical Supplier No. 1742350 MG
An active, estrogenic metabolite of daidzein; has vasodilatory action on rat isolated aortic rings at 1 µg/ml; stimulates the estrogen receptor-dependent growth of breast cancer MCF-7 cells at micromolar concentrations