Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrodaidzein 93.0+%, TCI America™
SDP

Catalog No. D423925MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D423925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D423925MG Supplier TCI America Supplier No. D423925MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17238-05-0
Molecular Formula C15H12O4
Molecular Weight (g/mol) 256.257
MDL Number MFCD00210494
InChI Key JHYXBPPMXZIHKG-UHFFFAOYSA-N
Synonym 7-Hydroxy-3-(4-hydroxyphenyl)-4-chromanone
PubChem CID 176907
ChEBI CHEBI:75842
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C1C(C(=O)C2=C(O1)C=C(C=C2)O)C3=CC=C(C=C3)O

Specifications

Melting Point 250°C
Color Yellow
Quantity 25 mg
Formula Weight 256.26
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dihydrodaidzein
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.