Learn More
Medchemexpress LLC Dihydrodiol-ibrutinib | 1654820-87-7 | 99.9% | 474.51 g/mol | C25H26N6O4 | 5 MG
SDP
Catalog No. 5000213654
Encompass_Preferred
Dihydrodiol-ibrutinib is a dihydrodiol active metabolite of ibrutinib that retains BTK inhibitory activity at a lower potency than the parent compound. Supplied as a white to off-white solid for laboratory research use, it is suitable for in vitro assays and formulation studies and can be prepared in common solvents for dosing formulations.
  • Active metabolite with measurable BTK inhibition (~15x lower than parent)
  • High reported purity, suitable for research applications
  • White to off-white solid for convenient handling
  • Soluble in DMSO and compatible with multiple in vivo formulation vehicles
  • Stable under recommended cold storage conditions for extended periods
Catalog No. 50-002-13654 Supplier Medchemexpress LLC Supplier No. HY1006595MG
Dihydrodiol-ibrutinib is a dihydrodiol active metabolite of ibrutinib that retains BTK inhibitory activity at a lower potency than the parent compound. Supplied as a white to off-white solid for laboratory research use, it is suitable for in vitro assays and formulation studies and can be prepared in common solvents for dosing formulations.
  • Active metabolite with measurable BTK inhibition (~15x lower than parent)
  • High reported purity, suitable for research applications
  • White to off-white solid for convenient handling
  • Soluble in DMSO and compatible with multiple in vivo formulation vehicles
  • Stable under recommended cold storage conditions for extended periods