Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydrodutasteride | 164656-22-8 | MFCD24386648 | 99.7% | C27H32F6N2O2 | 10MG
SDP

Catalog No. 5000259702
Encompass_Preferred

Dihydrodutasteride is a metabolite of dutasteride used as a research reagent for studies of 5α-reductase activity and dutasteride metabolism. It is supplied as a white to off-white solid intended for laboratory research use only; not for human or veterinary use.

  • High purity: 99.7% (LCMS)
  • Molecular formula C27H32F6N2O2 and molecular weight 530.55 g/mol
  • Appearance: white to off-white solid
  • Solubility ≥2.5 mg/mL (≈4.71 mM) in solvent
  • Intended for research use only, not for human or veterinary use

Catalog No. 50-002-59702 Supplier Medchemexpress LLC Supplier No. HY13538510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dihydrodutasteride is a metabolite of dutasteride used as a research reagent for studies of 5α-reductase activity and dutasteride metabolism. It is supplied as a white to off-white solid intended for laboratory research use only; not for human or veterinary use.

  • High purity: 99.7% (LCMS)
  • Molecular formula C27H32F6N2O2 and molecular weight 530.55 g/mol
  • Appearance: white to off-white solid
  • Solubility ≥2.5 mg/mL (≈4.71 mM) in solvent
  • Intended for research use only, not for human or veterinary use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.