Learn More
Medchemexpress LLC β-Dihydroequilin-d5 | 275.40 | 1 MG
SDP
Catalog No. 5000348597
Encompass_Preferred
β-Dihydroequilin-d5 is a deuterium-labeled version of β-Dihydroequilin, intended for research use only. Stable heavy isotopes such as deuterium are incorporated into drug molecules as tracers for quantitative analysis during drug development, with potential impact on pharmacokinetic and metabolic profiles.
  • Deuterium-labeled β-Dihydroequilin for research
  • Functions as a tracer for quantitative analysis in drug development
  • May influence pharmacokinetic and metabolic profiles of drugs
  • Stable under recommended storage conditions
  • Presented as a solid
Catalog No. 50-003-48597 Supplier Medchemexpress LLC Supplier No. HY138112S11MG
Add to Cart
Add to Cart
β-Dihydroequilin-d5 is a deuterium-labeled version of β-Dihydroequilin, intended for research use only. Stable heavy isotopes such as deuterium are incorporated into drug molecules as tracers for quantitative analysis during drug development, with potential impact on pharmacokinetic and metabolic profiles.
  • Deuterium-labeled β-Dihydroequilin for research
  • Functions as a tracer for quantitative analysis in drug development
  • May influence pharmacokinetic and metabolic profiles of drugs
  • Stable under recommended storage conditions
  • Presented as a solid