Learn More
Medchemexpress LLC Dihydroergocristine mesylate | 24730-10-7 | 1 ML
SDP
Catalog No. 5000420370
Encompass_Preferred
Dihydroergocristine mesylate (DHEC mesylate) is an inhibitor of γ-secretase (GSI) that effectively reduces the production of amyloid-β peptides associated with Alzheimer's disease. It directly binds to γ-secretase and Nicastrin, exhibiting high affinity. This product is for research use only.
  • Inhibits γ-secretase activity in T100 cells without affecting cell viability
  • Decreases cellular Aβ production
  • Causes dose-dependent accumulation of carboxy-terminal fragments of APP (APP-CTFs)
  • Reduces γ-secretase activity in fibroblast cells
Catalog No. 50-004-20370 Supplier Medchemexpress LLC Supplier No. HYN231910MM/1ML
Dihydroergocristine mesylate (DHEC mesylate) is an inhibitor of γ-secretase (GSI) that effectively reduces the production of amyloid-β peptides associated with Alzheimer's disease. It directly binds to γ-secretase and Nicastrin, exhibiting high affinity. This product is for research use only.
  • Inhibits γ-secretase activity in T100 cells without affecting cell viability
  • Decreases cellular Aβ production
  • Causes dose-dependent accumulation of carboxy-terminal fragments of APP (APP-CTFs)
  • Reduces γ-secretase activity in fibroblast cells