Learn More
Medchemexpress LLC Dihydroergocristine mesylate | 24730-10-7 | MFCD00153792 | ≥98.0% | 707.84 | C36H45N5O8S | 10 MG
SDP
Catalog No. 5000289627
Encompass_Preferred
Dihydroergocristine mesylate is a research reagent reported to act as a γ-secretase inhibitor; it reduces production of amyloid-β peptides and binds directly to γ-secretase and nicastrin. It is supplied as the mesylate salt for use in neuroscience and Alzheimer's disease research.
  • Acts as a γ-secretase inhibitor.
  • Reduces production of amyloid-β peptides.
  • Binds directly to γ-secretase and nicastrin.
  • Supplied as the mesylate salt.
  • Available in small research quantities suitable for analytical or in-vitro studies.
Catalog No. 50-002-89627 Supplier Medchemexpress LLC Supplier No. HYN231910MG
Dihydroergocristine mesylate is a research reagent reported to act as a γ-secretase inhibitor; it reduces production of amyloid-β peptides and binds directly to γ-secretase and nicastrin. It is supplied as the mesylate salt for use in neuroscience and Alzheimer's disease research.
  • Acts as a γ-secretase inhibitor.
  • Reduces production of amyloid-β peptides.
  • Binds directly to γ-secretase and nicastrin.
  • Supplied as the mesylate salt.
  • Available in small research quantities suitable for analytical or in-vitro studies.