Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydroergocristine methanesulfonate, Thermo Scientific Chemicals

Catalog No. AAJ60638EX3
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60638EX3 200 mg
AAJ6063803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60638EX3 Supplier Thermo Scientific Chemicals Supplier No. J60638EX3
Add to Cart

missing translation for 'validMainframeMsg'

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24730-10-7
Molecular Formula C36H45N5O8S
Molecular Weight (g/mol) 707.843
MDL Number MFCD00153792
InChI Key SPXACGZWWVIDGR-SPZWACKZSA-N
Synonym dihydroergocristine mesylate, dihydroergocristinmesilat, unii-ds7cl18uam, dihydroergocristine mesilate, 9,10-dihydroergocristine methanesulfonate, ds7cl18uam, dihydroergocristine methanesulfonate, dihydroergocristine methanesulfonate salt, 5'alpha-benzyl-9,10alpha-dihydro-12'-hydroxy-2'-isopropylergotaman-3',6',18-trione monomethanesulphonate, diertine tn
PubChem CID 444034
ChEBI CHEBI:31490
SMILES CC(C)C1(C(=O)N2C(C(=O)N3CCCC3C2(O1)O)CC4=CC=CC=C4)NC(=O)C5CC6C(CC7=CNC8=CC=CC6=C78)N(C5)C.CS(=O)(=O)O

Specifications

Color White
Quantity 200 mg
Merck Index 14,3653
Formula Weight 707.8
Physical Form Powder
Chemical Name or Material Dihydroergocristine methanesulfonate
EINECSNumber 246-434-6
RTECSNumber KE7600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.