Learn More
Medchemexpress LLC Dihydroergotamine mesylate | 6190-39-2 | 99.9% | 25 MG
SDP
Catalog No. 5000450667
Encompass_Preferred
Dihydroergotamine mesylate is a BBB-permeable ergot alkaloid primarily used for the research of migraines.
  • Decreases cell survival in A549 and NCI-H226 cells.
  • Inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells.
  • Induces mitochondrial dysfunction and apoptosis in A549 cells.
  • Inhibits STAT3 activation and enhances Mcl-1 protein stability.
  • Reduces tumor growth in xenografted mouse models when combined with other agents.
Catalog No. 50-004-50667 Supplier Medchemexpress LLC Supplier No. 25MGHYB0670A
Dihydroergotamine mesylate is a BBB-permeable ergot alkaloid primarily used for the research of migraines.
  • Decreases cell survival in A549 and NCI-H226 cells.
  • Inhibits cell survival in Hep3B, PLC/PRF/5, Huh7, and HepG2 cells.
  • Induces mitochondrial dysfunction and apoptosis in A549 cells.
  • Inhibits STAT3 activation and enhances Mcl-1 protein stability.
  • Reduces tumor growth in xenografted mouse models when combined with other agents.